missing translation for 'onlineSavingsMsg'
Learn More

DMRTC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DMRTC1B
Concentration 0.24 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16871065
View Documents
Proteintech
21473-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16861065
View Documents
Proteintech
21473-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DMRTC1B, also named as Doublesex- and mab-3-related transcription factor C1, is a 192 amino acid protein, which belongs to the DMRT family. DMRTC1B is predomitly expressed in kidney, pancreas, ovary and testis. Detected in brain and in many other tissues. DMRTC1B exists two isoform and molecular weight of isoforms are 20 kDa and 13 kDa.
Specifications

Specifications

DMRTC1B
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q5HYR2
632708, 728656
DMRTC1B Fusion Protein Ag15419
Primary
-20°C
DMRTC1B
0.24 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DMRT like family C1B, DMRTC1, DMRTC1B
Dmrtc1b
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.