missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRTA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DMRTA1 |
|---|---|
| Concentration | 0.65 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
May be involved in sexual development.Specifications
| DMRTA1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q5VZB9, Q8CFG4 | |
| 242523, 313352, 63951 | |
| DMRTA1 Fusion Protein Ag18796 | |
| Primary | |
| -20°C | |
| DMRTA1 |
| 0.65 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DMO, DMRT like family A1, DMRTA1 | |
| Dmrta1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title