missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRTA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92155-0.02ml
This item is not returnable.
View return policy
Description
DMRTA1 Polyclonal antibody specifically detects DMRTA1 in Mouse samples. It is validated for Western Blot
Specifications
| DMRTA1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DMO, DMRT-like family A1, doublesex- and mab-3-related transcription factor A1, MGC163307, MGC163309 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 205-504 of human DMRTA1 (NP_071443.2). SATPAFEVFQQDYPEEKQEQKESKCESCQNGQEELISKSHQLYLGSSSRSNGVIGKQSIGSSISEYSNKPDSILSPHPGEQSGGEESPRSLSSSDLESGNESEWVKDLTATKASLPTVSSRPRDPLDILTKIFPNYRRSRLEGILRFCKGDVVQAIEQVLNGKEHKPDNRNLANSEELENTAFQRASSFSLAGIGFGTLGNKSAFSPLQTTSASYGGDSSLYGVNPRVGISPLRLAYSSAGRGLSGFMSPYLTPGLVPTLPFRPALDYAFSGMIRDSSYLSSKDSITCGRLYFRPNQDNP | |
| 0.02 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 63951 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction