missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT2 Mouse anti-Human, Clone: PCRP-DMRT2-1B11, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Especificaciones
| Antigen | DMRT2 |
|---|---|
| Clone | PCRP-DMRT2-1B11 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
18305644
|
Novus Biologicals
NBP3-13840-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18337875
|
Novus Biologicals
NBP3-13840-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein ArrayEspecificaciones
| DMRT2 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y5R5 | |
| 10655 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-DMRT2-1B11 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein | |
| Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5) | |
| Primary | |
| Store at 4°C. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto