missing translation for 'onlineSavingsMsg'
Learn More

DMRT2 Antibody (PCRP-DMRT2-1B11), PE, Novus Biologicals™

Product Code. 18357524 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357524 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357524 Supplier Novus Biologicals Supplier No. NBP314270PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT2
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-DMRT2-1B11
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein
Host Species Mouse
Immunogen Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10655
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.