missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMRT1 Monoclonal Antibody (OTI4E12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526903
This item is not returnable.
View return policy
Description
DMRT1 Monoclonal Antibody for Western Blot, IHC (P)
This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.
Specifications
| DMRT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y5R6 | |
| DMRT1 | |
| Human recombinant protein fragment corresponding to amino acids 1-210 of DMRT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E12 | |
| Unconjugated | |
| DMRT1 | |
| CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119) | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1761 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction