missing translation for 'onlineSavingsMsg'
Learn More

DMPQ dihydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18033902
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18033902

Brand: Tocris Bioscience 1222/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, selective inhibitor of PDGFRβ

A potent and selective inhibitor of human vascular β-type platelet derived growth factor receptor tyrosine kinase (PDGFRβ) (IC50 = 80 nM). Displays > 100-fold selectivity over EGFR tyrosine kinase, erbB2, p56, protein kinase A and protein kinase C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123491-15-5
Molecular Formula C16H16Cl2N2O2
Molecular Weight (g/mol) 339.216
InChI Key YBBAOKYVJCNJIV-UHFFFAOYSA-N
Synonym dmpq dihydrochloride, 5,7-dimethoxy-3-4-pyridinyl quinoline dihydrochloride, 5,7-dimethoxy-3-pyridin-4-yl quinoline dihydrochloride, dmpq hydrochloride, dmpq dihydrochloride hplc, 5,7-dimethoxy-3-4-pyridinyl quinolinedihydrochloride
PubChem CID 45073427
IUPAC Name 5,7-dimethoxy-3-pyridin-4-ylquinoline;dihydrochloride
SMILES COC1=CC2=NC=C(C=C2C(=C1)OC)C3=CC=NC=C3.Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 339.22
Percent Purity >99%
Chemical Name or Material DMPQ dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.