missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMPK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542121
This item is not returnable.
View return policy
Description
Peptide sequence: TRLGRGGAGD FRTHPFFFGL DWDGLRDSVP PFTPDFEGAT DTCNFDLVED Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues. Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy. The protein encoded by this gene is a serine-threonine kinase that is closely related to other kinases that interact with members of the Rho family of small GTPases. Substrates for this enzyme include myogenin, the beta-subunit of the L-type calcium channels, and phospholemman. The 3' untranslated region of this gene contains 5-37 copies of a CTG trinucleotide repeat. Expansion of this unstable motif to 50-5,000 copies causes myotonic dystrophy type I, which increases in severity with increasing repeat element copy number. Repeat expansion is associated with condensation of local chromatin structure that disrupts the expression of genes in this region.
Specifications
| DMPK | |
| Polyclonal | |
| Unconjugated | |
| DMPK | |
| DM; DM protein kinase; DM1; DM1 protein kinase; Dm15; DM1PK; DMK; DM-kinase; Dmpk; dystrophia myotonica kinase, B15; dystrophia myotonica protein kinase; dystrophia myotonica-protein kinase; MDPK; MT-PK; myotonic dystrophy associated protein kinase; myotonic dystrophy protein kinase; myotonin protein kinase A; myotonin-protein kinase; thymopoietin homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q09013 | |
| DMPK | |
| Synthetic peptide directed towards the middle region of human DMPK. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction