missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524463
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
Calcium-binding protein which may play a role in dentin mineralization.
Specifications
| DMP4 | |
| Polyclonal | |
| Unconjugated | |
| FAM20C | |
| C76981; Dentin matrix protein 4; DMP4; DMP-4; Extracellular serine/threonine protein kinase FAM20C; Fam20c; family with sequence similarity 20 member C; family with sequence similarity 20, member C; GEF-CK; Golgi casein kinase; golgi-enriched fraction casein kinase; Kiaa4081; mKIAA4081; RGD1311980; RNS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IXL6 | |
| FAM20C | |
| KLH conjugated synthetic peptide between 443-471 amino acids from the C-terminal region of human DMP4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction