Learn More
DMP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA588069
Description
Immunogen sequence: SGGSQLLEHP SRKIFRKSRI SEEDDRSELD DNNTMEEVKS DSTENSNSRD TGLSQPRRDS KGDSQEDSKE NLSQEESQNV DGPSSESSQE ANLSSQENSS ESQEEVVSES RGDNPDPTTS YVEDQEDSDS SEEDSSHTLS HSKSESREEQ ADSESSESLN FSEESPESPE DENSSSQEGL QSHSSSAESQ SEESHSEEDD SDSQDSSRSK EDSNSTESKS SSEEDGQLKN IEIESRKLTV DAYHNKPIGD QDDNDCQDGY; Positive Samples: U-937, U-251MG, Mouse brain; Cellular Location: Cytoplasm, Nucleus, extracellular matrix, extracellular space
Dentin matrix acidic phosphoprotein is an extracellular matrix protein and a member of the small integrin binding ligand N-linked glycoprotein family. This protein, which is critical for proper mineralization of bone and dentin, is present in diverse cells of bone and tooth tissues. The protein contains a large number of acidic domains, multiple phosphorylation sites, a functional arg-gly-asp cell attachment sequence, and a DNA binding domain. In undifferentiated osteoblasts it is primarily a nuclear protein that regulates the expression of osteoblast-specific genes. During osteoblast maturation the protein becomes phosphorylated and is exported to the extracellular matrix, where it orchestrates mineralized matrix formation. Mutations in the gene are known to cause autosomal recessive hypophosphatemia, a disease that manifests as rickets and osteomalacia. The gene structure is conserved in mammals. Two transcript variants encoding different isoforms have been described for this gene.
Specifications
| DMP1 | |
| Polyclonal | |
| Unconjugated | |
| DMP1 | |
| Dmp1 | |
| Recombit fusion protein containing a sequence corresponding to amino acids 254-513 of human DMP1 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunofluorescence, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O55188, Q13316 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13406, 1758 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.