missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMP1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547621
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human DMP-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Dentin matrix acidic phosphoprotein is an extracellular matrix protein and a member of the small integrin binding ligand N-linked glycoprotein family. This protein, which is critical for proper mineralization of bone and dentin, is present in diverse cells of bone and tooth tissues. The protein contains a large number of acidic domains, multiple phosphorylation sites, a functional arg-gly-asp cell attachment sequence, and a DNA binding domain. In undifferentiated osteoblasts it is primarily a nuclear protein that regulates the expression of osteoblast-specific genes. During osteoblast maturation the protein becomes phosphorylated and is exported to the extracellular matrix, where it orchestrates mineralized matrix formation. Mutations in the gene are known to cause autosomal recessive hypophosphatemia, a disease that manifests as rickets and osteomalacia. The gene structure is conserved in mammals. Two transcript variants encoding different isoforms have been described for this gene.
Specifications
| DMP1 | |
| Polyclonal | |
| Unconjugated | |
| Dmp1 | |
| AG1; ARHP; ARHR; AV020965; dentin matrix acidic phosphoprotein 1; dentin matrix protein 1; DENTMAT; Dmp; DMP 1; Dmp1; DMP-1; MGC130441; PP; serine rich acidic phosphoprotein | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 13406 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O55188 | |
| Dmp1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DMP-1 Leu17-Tyr503. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction