missing translation for 'onlineSavingsMsg'
Learn More

DMP 777, MedChemExpress

Product Code. 16490638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490638 5 mg 5mg
16484097 50 mg 50mg
16408208 10 mg 10mg
16403776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490638 Supplier Medchem Express Supplier No. HY75957
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.

DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.

Specifications

Chemical Name or Material DMP 777
CAS 157341-41-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H40N4O6
Quantity 5 mg
Synonym L-694458
Solubility Information DMSO : 38.33 mg/mL (67.88 mM; Need ultrasonic)
SMILES CCC[C@H](C1=CC2=C(C=C1)OCO2)NC(N3[C@H](C(CC)(C3=O)CC)OC4=CC=C(C=C4)C(N5CCN(CC5)C)=O)=O
Molecular Weight (g/mol) 564.67
Formula Weight 564.67
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.