missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DMP-1 Polyclonal specifically detects DMP-1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | DMP-1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP-1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant DMP-1 (Q13316-1; Met 1-Tyr 513). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?