missing translation for 'onlineSavingsMsg'
Learn More

DMOG, MedChemExpress

Product Code. 16452967
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16452967

Brand: Medchem Express HY15893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal. DMOG induces cell autophagy.

DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal. DMOG induces cell autophagy.

Specifications

Chemical Name or Material DMOG
CAS 89464-63-1
Color White
Purity Grade Notes Research
Molecular Formula C6H9NO5
Quantity 50 mg
Synonym Dimethyloxallyl Glycine
Solubility Information DMSO : ≥ 50 mg/mL (285.49 mM) ∣H2O : 50 mg/mL (285.49 mM; Need ultrasonic)
SMILES O=C(OC)CNC(C(OC)=O)=O
Molecular Weight (g/mol) 175.14
Formula Weight 175.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.