missing translation for 'onlineSavingsMsg'
Learn More

DMNQ, MedChemExpress

Product Code. 16497717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497717 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497717 Supplier Medchem Express Supplier No. HY121026
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DMNQ is a redox cycling agent that generates both superoxide and hydrogen peroxide intracellularly in a concentration dependent manner. DMNQ increases ROS generation.

DMNQ is a redox cycling agent that generates both superoxide and hydrogen peroxide intracellularly in a concentration dependent manner. DMNQ increases ROS generation.

Specifications

Chemical Name or Material DMNQ
CAS 6956-96-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H10O4
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (229.14 mM; Need ultrasonic)
SMILES O=C1C(OC)=C(C(C2=CC=CC=C21)=O)OC
Molecular Weight (g/mol) 218.21
Formula Weight 218.21
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.