missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DMH2

Product Code. 18770128
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770128 50 mg 50mg
18716158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18770128 Supplier Tocris Bioscience™ Supplier No. 5580/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Type I BMP receptor inhibitor

DMH2 is a type I BMP receptor inhibitor (Ki values are <1, 5.4 and 43 nM for ALK6, ALK3 and ALK2, respectively). Exhibits >30-fold selectivity over ALK4, ALK5, BMPR2, AMPK and VEGFR2. Also binds to TGFBR2 (Ki = 85 nM). Decreases growth and induces cell death of H1299 non-small cell lung carcinoma cells in vitro. Enhances liver regeneration in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[6-[4-[2-(4-Morpholinyl)ethoxy]phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline
CAS 1206711-14-9
Quantity 50 mg
Target BMP and Other Activin Receptor Inhibitors
Molecular Formula C27H25N5O2
Formula Weight 451.52
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.