missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies DMH1

Product Code. p-7217396
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17140252

Brand: STEMCELL Technologies 73632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activin/Nodal/TGFβ pathway inhibitor; Inhibits ALK2

  • DMH1 (dorsomorphin homolog 1) is a selective inhibitor of activin receptor-like kinase 2 (ALK2; IC50 = 13 - 108 nM), a type I bone morphogenetic protein (BMP) receptor (Hao et al.; Mohedas et al.) DMH1 exhibits no detectable inhibition of ALK4, ALK5, AMPK, KDR (VEGFR2), or PDGFRβ, although it inhibits ALK1 and ALK3 at nanomolar concentrations (Hao et al.; Mohedas et al.)

Specifications

CAS 1206711-16-1
Synonym Dorsomorphin homolog 1
Quantity 1 mg
Molecular Formula C24H20N4O
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.