missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24813-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. Alternative splicing results in multiple transcript variants.Specifications
| DMGDH | |
| Polyclonal | |
| Unconjugated | |
| DMGDH | |
| dimethylglycine dehydrogenase, DMGDH, DMGDHD, M2GD, ME2GLYDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245961, 29958, 74129 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q63342, Q9DBT9, Q9UI17 | |
| Dmgdh | |
| DMGDH Fusion Protein Ag20529 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction