missing translation for 'onlineSavingsMsg'
Learn More

DMEQ-TAD, TRC

Product Code. 30381102
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381102 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381102 Supplier Toronto Research Chemicals Supplier No. TRCD49440025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DMEQ-TAD
CAS 132788-52-4
Molecular Formula C15 H15 N5 O5
Quantity 25 mg
Synonym 4-[2-(3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinyl)ethyl]-3H-1,2,4-triazole-3,5(4H)-dione
InChI Formula InChI=1S/C15H15N5O5/c1-19-10-7-12(25-3)11(24-2)6-9(10)16-8(13(19)21)4-5-20-14(22)17-18-15(20)23/h6-7H,4-5H2,1-3H3
SMILES COc1cc2N=C(CCN3C(=O)N=NC3=O)C(=O)N(C)c2cc1OC
IUPAC Name 4-[2-(6,7-dimethoxy-4-methyl-3-oxo-quinoxalin-2-yl)ethyl]-1,2,4-triazole-3,5-dione
Molecular Weight (g/mol) 345.31
Formula Weight 345.1073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.