Learn More
MP Biomedicals™ DMEM Powder with 4500 mg/L Dextrose, L-Glutamine, without Sodium Bicarbonate
Shop All MP Biomedicals ProductsDescription
Dulbecco's Modified Eagle's Medium (DMEM) is the most widely used modification of Eagle's Basal Medium (BME). DMEM contains four times greater concentration of amino acids, vitamins and supplementary components, and is ideally suited for supporting and maintaining a vast range of mammalian cell types. The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures. For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available.
Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Specifications
Specifications
| Product Type | Cell Culture Media |
| With Additives | Dextrose, Glutamine |
| Without Additives | No Sodium Bicarbonate |
| Glucose Concentration | High Glucose |
| Formulation | Powder to Make 5 L Solution |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.