missing translation for 'onlineSavingsMsg'
Learn More

Sartorius DMEM High Glucose without L-Glutamine

Product Code. 31908442 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
1mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908442 2 x 500 mL 1mL
31908389 5 x 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31908442 Supplier Sartorius Supplier No. 010559A
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

  • Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells
  • Offers a variety of DMEM modifications for a range of cell culture applications

Specifications

Classification Serum-free
Form Liquid
Product Type DMEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glucose, Sodium Bicarbonate
Without Additives No Glutamine
Glucose Concentration 2X
Quantity 2 x 500 mL
Culture Type Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.