missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM (Dulbecco’s Modified Eagle’s Medium)
BIOPHARMA

Product Code. 15333601
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15333601 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15333601 Supplier Corning™ Supplier No. 10027CV
  • ONLINE EXCLUSIVE PRICE
    £176.75
    £156.90 / Pack of 6
    Save £19.85
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME

  • Low endotoxin - <0.25 EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
TRUSTED_SUSTAINABILITY

Specifications

Product Type DMEM Media
Quantity 500 mL
Endotoxin Level <0.25 EU/mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.