missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies DMEM with 4500 mg/L D-Glucose

Product Code. 17118091
Encompass
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17118091

Brand: STEMCELL Technologies 36250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dulbecco's Modified Eagle's Medium (DMEM) with 4500 mg/L D-glucose

  • Dulbecco's Modified Eagle's Medium (DMEM) with 4500 mg/L D-­Glucose is recommended for a wide variety of cell culture applications
  • Selection of suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition required for the cell culture application

Specifications

For Use With (Application) Cell Culture
Quantity 500 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.