missing translation for 'onlineSavingsMsg'
Learn More

DMCM hydrochloride, MedChemExpress

Product Code. 16485707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485707 5 mg 5mg
16479536 50 mg 50mg
16464286 100 mg 100mg
16462968 10 mg 10mg
16412566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485707 Supplier Medchem Express Supplier No. HY100369A
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DMCM hydrochloride is a nonselective full inverse agonist of benzodiazepine. DMCM shows bnding afinity at human recombinant GABAA αxβ3γ2 receptor subtypes with Kis of 10 nM, 13 nM, 7.5 nM, 2.2 nM for α1, α2, α3, and α5 receptors, respectively.

DMCM hydrochloride is a nonselective full inverse agonist of benzodiazepine. DMCM shows bnding afinity at human recombinant GABAA αxβ3γ2 receptor subtypes with Kis of 10 nM, 13 nM, 7.5 nM, 2.2 nM for α1, α2, α3, and α5 receptors, respectively.

Specifications

Chemical Name or Material DMCM hydrochloride
CAS 1215833-62-7
Color Green
Purity Grade Notes Research
Molecular Formula C17H19ClN2O4
Quantity 5 mg
Solubility Information H2O : 25 mg/mL (71.27 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (28.51 mM; Need ultrasonic)
SMILES O=C(C1=C(CC)C2=C(C=N1)NC3=C2C=C(OC)C(OC)=C3)OC.[H]Cl
Molecular Weight (g/mol) 350.8
Formula Weight 350.8
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.