missing translation for 'onlineSavingsMsg'
Learn More

DMCM hydrochloride, MedChemExpress

Product Code. 16479536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479536

Brand: Medchem Express HY100369A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DMCM hydrochloride is a nonselective full inverse agonist of benzodiazepine. DMCM shows bnding afinity at human recombinant GABAA αxβ3γ2 receptor subtypes with Kis of 10 nM, 13 nM, 7.5 nM, 2.2 nM for α1, α2, α3, and α5 receptors, respectively.

DMCM hydrochloride is a nonselective full inverse agonist of benzodiazepine. DMCM shows bnding afinity at human recombinant GABAA αxβ3γ2 receptor subtypes with Kis of 10 nM, 13 nM, 7.5 nM, 2.2 nM for α1, α2, α3, and α5 receptors, respectively.

Specifications

Chemical Name or Material DMCM hydrochloride
CAS 1215833-62-7
Color Green
Purity Grade Notes Research
Molecular Formula C17H19ClN2O4
Quantity 50 mg
Solubility Information H2O : 25 mg/mL (71.27 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (28.51 mM; Need ultrasonic)
SMILES O=C(C1=C(CC)C2=C(C=N1)NC3=C2C=C(OC)C(OC)=C3)OC.[H]Cl
Molecular Weight (g/mol) 350.8
Formula Weight 350.8
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.