missing translation for 'onlineSavingsMsg'
Learn More

DMC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7206295 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16822074 20 μL 20µL
16812074 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16822074 Supplier Proteintech Supplier No. 137141AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.

Specifications

Antigen DMC1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DMC1
Gene Accession No. Q14565, Q61880
Gene Alias DMC1, DMC1H, HsLim15, LIM15
Gene Symbols Dmc1
Host Species Rabbit
Immunogen DMC1 Fusion Protein Ag4653
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11144, 13404, 362960
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.