missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMC1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13228868
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13228868 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13228868 Supplier Invitrogen™ Supplier No. PA521472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa, Molt-4, Raji, mouse testis. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (83%), Japanese Medaka (84%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen DMC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DMC1
Gene Accession No. Q14565, Q61880
Gene Alias DCM1; disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; dJ199H16.1; DMC 1; DMC1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; DMC1H; DNA meiotic recombinase 1; HsLim15; LIM15; Mei11; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4; sgdp
Gene Symbols DMC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DMC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11144, 13404
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.