missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMC1 Monoclonal Antibody (2H12/4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA120220
This item is not returnable.
View return policy
Description
MA1-20220 detects DMC1 in human, mouse, rat, and bovine samples.
DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
Specifications
| DMC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q14565, Q61880 | |
| DMC1 | |
| Recombinant fusion protein (Human). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2H12/4 | |
| Unconjugated | |
| DMC1 | |
| DCM1; disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; dJ199H16.1; DMC 1; DMC1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; DMC1H; DNA meiotic recombinase 1; HsLim15; LIM15; Mei11; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4; sgdp | |
| Mouse | |
| Protein A | |
| RUO | |
| 11144, 13404, 362960, 618398 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction