missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMC1 Antibody (2H12/4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2617C
This item is not returnable.
View return policy
Description
DMC1 Monoclonal antibody specifically detects DMC1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
| DMC1 | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DMC1 | |
| Human DMC1 protein (expressed as 6xHis fusion in E. coli and band cut out of gel of insoluble pelet) | |
| Primary | |
| Specific for DMC1 - does not cross-react with the related protein Rad51. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2H12/4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473 | |
| Mouse | |
| 0.1 mL | |
| 11144 | |
| Human, Mouse, Bovine, Bovine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction