missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMBX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555425
This item is not returnable.
View return policy
Description
Immunogen sequence: SLTITPVAPG GGLLGPSHSY SSSPLSLFRL QEQFRQHMAA TNNLVHYSSF EVGGPAPAAA AAAAAVPYLG VNMAPLGSLH CQSYYQSLSA AAAAHQGVWG SPLLPAPPAG LAPASATLNS KTTSIENLRL RAKQHAASLG LDTLP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| DMBX1 | |
| Polyclonal | |
| Unconjugated | |
| DMBX1 | |
| Atx; Cdmx; diencephalon/mesencephalon homeobox 1; diencephalon/mesencephalon homeobox protein 1; Diencephalon/mesencephalon-expressed brain homeobox gene 1 protein; dmbx1; homeobox gene Atx; homeoprotein MBX; MBX; Orthodenticle homolog 3; OTX; OTX3; Paired-like homeobox protein DMBX1; paired-type homeobox Atx; PaxB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFW5 | |
| DMBX1 | |
| Recombinant protein corresponding to Human DMBX1. Recombinant protein control fragment (Product #RP-89442). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction