missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMBT1/GP340 Antibody (548031) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5915JF635
This item is not returnable.
View return policy
Description
DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| DMBT1/GP340 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein | |
| E. Coli-derived recombinant mouse DMBT1, Trp215-Gly420, Accession # Q60997 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 548031 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Breast Cancer | |
| 1755 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction