missing translation for 'onlineSavingsMsg'
Learn More

DMAP1 Antibody, HRP, Novus Biologicals™

Product Code. 18414899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414899 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414899 Supplier Novus Biologicals Supplier No. NB100256H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DMAP1 Polyclonal antibody specifically detects DMAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMAP1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias DNA methyltransferase 1 associated protein 1, DNA methyltransferase 1-associated protein 1, DNMAP1DNMT1-associated protein 1, DNMT1 associated protein 1, DNMTAP1, EAF2, FLJ11543, KIAA1425DKFZp686L09142, MEAF2, SWC4
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human DNA Methyltransferase 1 Associated Protein 1 using the numbering given in entry NP_061973.1 (GeneID 55929).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, DNA Repair, MAP Kinase Signaling
Primary or Secondary Primary
Gene ID (Entrez) 55929
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.