missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DMA trihydrochloride

Product Code. 16499247
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499247 5 mg 5mg
16456846 100 mg 100mg
16442777 50 mg 50mg
16426698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499247 Supplier MedChemExpress Supplier No. HY15621A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DMA trihydrochloride is a fluorescent compound (λex=340 nm, λem=478 nm).

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : 15.9 mg/mL (27.51 mM; Need ultrasonic and warming)
Molecular Weight (g/mol) 577.93 g/mol
CAS 2095832-33-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.