missing translation for 'onlineSavingsMsg'
Learn More

DM4, MedChemExpress

Product Code. 16477897
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477897

missing translation for 'mfr': Medchem Express HY12454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DM4 is is an antitubulin agent that inhibit cell division. DM4 can be used in the preparation of antibody drug conjugate.

DM4 is is an antitubulin agent that inhibit cell division. DM4 can be used in the preparation of antibody drug conjugate.

Specifications

Chemical Name or Material DM4
CAS 796073-69-3
Color White
Purity Grade Notes Research
Molecular Formula C38H54ClN3O10S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (128.14 mM; Need ultrasonic)
SMILES C[C@]1([C@H](CC(N(C(C=C2C=C3OC)=C3Cl)C)=O)OC([C@H](C)N(C)C(CCC(C)(S)C)=O)=O)[C@H]([C@@H]([C@](OC4=O)([H])C[C@]([C@](/C=C/C=C(C)/C2)([H])OC)(N4)O)C)O1
Molecular Weight (g/mol) 780.37
Formula Weight 780.37
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.