missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLX5 Monoclonal Antibody (712216)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524333
This item is not returnable.
View return policy
Description
In direct ELISAs, no cross-reactivity with recombinant human DLX4 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of a homeobox transcription factor gene family similar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation.
Specifications
| DLX5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P56178 | |
| DLX5 | |
| E. coli-derived recombinant human DLX5 Asn216-Tyr289. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 712216 | |
| Unconjugated | |
| DLX5 | |
| AI385752; distal-less homeo box 5; distal-less homeobox 5; distal-less homeobox protein 5; DLX-3; DLX5; Homeobox protein DLX-3; homeobox protein DLX-5; RDLX; SHFM1D; split hand/foot malformation type 1 with sensorineural hearing loss; Unknown (protein for MGC:137934) | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1749 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction