missing translation for 'onlineSavingsMsg'
Learn More

DLX5 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30732526 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732526 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30732526 Leverantör Novus Biologicals Leverantörsnummer AF6710JF669

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Sheep Polyclonal Antibody

DLX5 Polyclonal antibody specifically detects DLX5 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen DLX5
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias distal-less homeo box 5, distal-less homeobox 5, homeobox protein DLX-5
Host Species Sheep
Immunogen E. coli-derived recombinant human DLX5, Asn216-Tyr289, Accession # P56178
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1749
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.