missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DLX3 Recombinant Rabbit Monoclonal Antibody (JE64-45)
GREENER_CHOICE

Product Code. 30276359
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276359 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30276359 Supplier Invitrogen™ Supplier No. MA556559

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DLX3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The distal-less homeobox 3 (DLX3) protein is a putative transcriptional activator that has been implicated during development and differentiation of epithelial tissues. This is a homeodomain transcription factor in vertebrates, related to Distal-less in Drosophila, which is expressed in differentiating epidermal cells, in neural crest, hair follicles, dental epithelium, mesenchyme, the optic and olfactory placodes, limb bud, placenta, and in the cement gland, which is located in the extreme anterior neural plate in Xenopus embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Dlx3
Gene Accession No. O60479
Gene Alias AI4; AV237891; DII C; distal-less homeobox 3; Dlx3; Dlx-3; Homeobox protein DLX-3; LOW QUALITY PROTEIN: homeobox protein DLX-3; TDO
Gene Symbols Dlx3
Host Species Rabbit
Immunogen Synthetic peptide within human dlx3 aa 238-287/287.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1747
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.