missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13261-3-AP-150UL
This item is not returnable.
View return policy
Description
The distal-less homeobox 3 (DLX3) protein is a putative transcriptional activator that has been implicated during development and differentiation of epithelial tissues. This is a homeodomain transcription factor in vertebrates, related to Distal-less in Drosophila, which is expressed in differentiating epidermal cells, in neural crest, hair follicles, dental epithelium, mesenchyme, the optic and olfactory placodes, limb bud, placenta, and in the cement gland, which is located in the extreme anterior neural plate in Xenopus embryos.Specifications
| DLX3 | |
| Polyclonal | |
| Unconjugated | |
| DLX3 | |
| AI4, distal less homeobox 3, DLX3, Homeobox protein DLX 3, TDO | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13393, 1747, 287638 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60479, Q64205 | |
| Dlx3 | |
| DLX3 Fusion Protein Ag3865 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction