missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109638
This item is not returnable.
View return policy
Description
Immunogen sequence: PAKAKPAEAP AAAAPKAEPT AAAVPPPAAP IPTQMPPVPS PSQPPSGKPV SAVKPTVAPP LAEPGAGKGL RSEHREKMNR MRQRIAQRLK EAQNTCAMLT T.
This gene encodes a mitochondrial protein that belongs to the 2-oxoacid dehydrogenase family. This protein is one of the three components (the E2 component) of the 2-oxoglutarate dehydrogenase complex that catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). Alternatively spliced transcript variants have been found for this gene.
Specifications
| DLST | |
| Polyclonal | |
| Unconjugated | |
| Dlst | |
| 1600017E01Rik; 2-oxoglutarate dehydrogenase complex component E2; 4632413C10Rik; 4930529O08Rik; dihydrolipoamide S-succinyltransferase; dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex); dihydrolipoamide succinyltransferase component of 2-oxoglutarate dehydrogenase complex; Dihydrolipoyllysine-residue succinyltransferase; dihydrolipoyllysine-residue succinyltransferase component of 2-oxoglutarate dehydrogenase complex, mitochondrial; DLST; DLTS; E2; E2K; OGDC-E2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1743, 299201, 78920 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.77 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P36957, Q01205, Q9D2G2 | |
| Dlst | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human DLST (NP_001924.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction