missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLST Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38349-100ul
This item is not returnable.
View return policy
Description
DLST Polyclonal antibody specifically detects DLST in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DLST | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 2-oxoglutarate dehydrogenase complex component E2, dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutaratecomplex), Dihydrolipoamide succinyltransferase component of 2-oxoglutarate dehydrogenasecomplex, dihydrolipoyllysine-residue succinyltransferase component of 2-oxoglutaratedehydrogenase complex, mitochondrial, DLTS, E2K, EC 2.3.1, EC 2.3.1.61, OGDC-E2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human DLST (NP_001924.2).,, Sequence:, MLSRSRCVSRAFSRSLSAFQKGNCPLGRRSLPGVSLCQGPGYPNSRKVVINNSVFSVRFFRTTAVCKDDLVTVKTPAFAESVTEGDVRWEKAVGDTVAEDEVVCEIETDKTSVQVPSPANGVIEALLVPDGGKVEGGTPLFTLRKTGAAPAKAKPAEAPAAAAPKAEPTAAAVPPPAAPIPTQMPPVPSPSQPPSGKPVSAVKPTVAPPLAEPGAGKGLRSEHREKMNRMRQRIAQRLKEAQNTCAMLTTFNEIDMSNIQEMRARHKEAFLKKHNLKLGFMSAFVKASAF | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1743 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction