Learn More
DLL4 (delta-like 4) Monoclonal Antibody (HMD4-1), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5948-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: This HMD4-1 monoclonal antibody reacts with the extracellular domain of mouse Delta-like 4, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g., Notch 1-4), Delta-like 4 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Delta-like 4 is expressed by thymic lymphoid and stromal cells, as well as macrophages and dendritic cells in the spleen. This molecule is also expressed in non-immune cells, including vascular smooth muscle and endothelial cells. Delta-like 4 plays a role in T cell development, regulation of experimental autoimmune encephalomyelitis, and the formation of mature vasculature. The HMD4-1 monoclonal antibody has been reported to have blocking activity and crossreact to rat. Applications Reported: The HMD4-1 antibody has been reported for use in functional assays. Applications Tested: This HMD4-1 antibody has been tested by flow cytometric analysis of mouse Delta-like 4-transfected cells. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.Specifications
DLL4 (delta-like 4) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
D3ZHH1, Q9JI71 | |
Dll4 | |
Affinity chromatography | |
RUO | |
311332, 54485 | |
4° C | |
Liquid |
Functional Assay | |
HMD4-1 | |
Functional Grade | |
Dll4 | |
AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341 | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
For Research Use Only.