missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25535-1-AP-150UL
This item is not returnable.
View return policy
Description
Delta-like protein 3 is a ligand for the Notch signaling pathway. It inhibits primary neurogenesis. DLL3 plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. Defects in DLL3 are the cause of Spondylocostal dysostosis autosomal recessive type 1 (SCDO1). Mutations in DLL3 gene cause truncal shortening relative to their limbs, which leads to abdominal protrusion, abnormal spinal curvature and sometimes a plagiocephaly-torticollis sequence. It may be required to divert neurons along a specific differentiation pathway.Specifications
| DLL3 | |
| Polyclonal | |
| Unconjugated | |
| DLL3 | |
| delta like 3 (Drosophila), Delta like protein 3, Delta3, DLL3, Drosophila Delta homolog 3, SCDO1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10683, 13389 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88516, Q9NYJ7 | |
| DLL3 | |
| DLL3 Fusion Protein Ag21965 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction