Learn More
DLL3 Rabbit, anti-Human, CoraLite Plus 647, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL647-25535-100UL
Description
The DLL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DLL3. This antibody recognizes human, mouse antigen. The DLL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 647.
Specifications
| DLL3 | |
| 1000 μg/mL | |
| delta like 3 (Drosophila), Delta like protein 3, Delta3, DLL3, Drosophila Delta homolog 3, SCDO1 | |
| 100 μL | |
| 10683 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 647 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |