missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL3 Antibody (rovalpituzumab-MMAE), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28461CL1
This item is not returnable.
View return policy
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DLL3 | |
| Monoclonal | |
| CoraFluor 1 | |
| delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
| DLL3 | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience | |
| 10683 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| rovalpituzumab-MMAE | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction