missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL3 Antibody (DLL3/9361), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | DLL3 |
|---|---|
| Clone | DLL3/9361 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30293595
|
Novus Biologicals
NBP3-44583-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30290748
|
Novus Biologicals
NBP3-44583-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| DLL3 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
| Recombinant fragment (around aa400-618) of human DLL3 protein (Uniprot: Q9NYJ7). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| DLL3/9361 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 10683 | |
| IgG2c κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title