missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLL1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547207
This item is not returnable.
View return policy
Description
This antibody detects recombinant human DLL1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human DLL4 and recombinant mouse DLL4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
Specifications
| DLL1 | |
| Polyclonal | |
| Unconjugated | |
| DLL1 | |
| C-Delta-1; Delta; delta like canonical Notch ligand 1; delta like-1; DELTA1; delta-like 1; delta-like 1 (Drosophila); delta-like protein 1; DL1; Dll1; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1; H-Delta-1; LOW QUALITY PROTEIN: delta-like protein 1; OTTHUMP00000017690; UNQ146/PRO172 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 28514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O00548 | |
| DLL1 | |
| Mouse myeloma cell line NS0-derived recombinant human DLL1 Ser22-Gly540. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction