missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DLK2/EGFL9 Monoclonal antibody specifically detects DLK2/EGFL9 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | DLK2/EGFL9 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 908866R |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | delta-like 2 homolog (Drosophila), DLK-2, EGFL9, EGF-like protein 9, EGF-like-domain, multiple 9, Epidermal growth factor-like protein 9, MGC111055, MGC2487, protein delta homolog 2 |
| Host Species | Mouse |
| Immunogen | Human embryonic kidney cell line HEK-293-derived transfected with human DLK2/EGFL9, Asp27-Ser306, Accession # Q6UY11 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?