missing translation for 'onlineSavingsMsg'
Learn More

DLK-IN-1, MedChemExpress

Product Code. 16431077
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431077

Brand: Medchem Express HY114331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.

DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.

Specifications

Chemical Name or Material DLK-IN-1
CAS 1620574-24-4
Color White
Purity Grade Notes Research
Molecular Formula C20H24F3N5O2
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (147.60 mM; Need ultrasonic)
SMILES CC(N1C([C@@H]2[C@]3([H])[C@@]2([H])CN(C4COC4)C3)=CC(C5=CN=C(N)C(OC(F)(F)F)=C5)=N1)C
Molecular Weight (g/mol) 423.43
Formula Weight 423.43
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.