missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLGAP3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81713C
This item is not returnable.
View return policy
Description
DLGAP3 Polyclonal antibody specifically detects DLGAP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| DLGAP3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| DAP-3, discs, large (Drosophila) homolog-associated protein 3, disks large-associated protein 3, SAP90/PSD 95 associated protein 3, SAP90/PSD-95-associated protein 3 | |
| DLGAP3 antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human DLGAP3. The immunogen is located within the last 50 amino acids of DLGAP3. Amino Acid Squence: KERSLDSVDRQRQEARKR | |
| 0.1 mL | |
| Neuroscience | |
| 58512 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction