missing translation for 'onlineSavingsMsg'
Learn More

DLG5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DLG5
Concentration 0.29 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16814974
View Documents
Proteintech
15687-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16804974
View Documents
Proteintech
15687-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May play a role at the plasma membrane in the maintece of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
Specifications

Specifications

DLG5
Western Blot
Unconjugated
Rabbit
Mouse, Human
E9Q9R9, Q8TDM6
71228, 9231
DLG5 Fusion Protein Ag8194
Primary
-20°C
DLG5
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Discs large protein P dlg, Disks large homolog 5, DLG5, KIAA0583, LP DLG, P DLG5, PDLG, Placenta and prostate DLG
Dlg5
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.